Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127893.1 | NP_001121365.1 | 3 | 973 | ||
NM_020219.3 | NP_064604.2 | 3 | 973 | |||
GenBank mRNA | AK027157.1 | - | 3 | 690 | ||
AK291588.1 | - | 3 | 674 | |||
AY358394.1 | - | 3 | 703 | |||
BC083499.1 | - | 3 | 973 | 30390224 | ||
KJ851726.1 | - | 3 | 493 | |||
KJ851727.1 | - | 3 | 493 | |||
KJ851728.1 | - | 3 | 493 | |||
KJ851729.1 | - | 3 | 493 | |||
KJ851730.1 | - | 3 | 493 | |||
KJ851731.1 | - | 3 | 493 | |||
KJ851732.1 | - | 3 | 493 | |||
KJ851733.1 | - | 3 | 493 | |||
KJ851734.1 | - | 3 | 493 | |||
KJ851735.1 | - | 3 | 493 | |||
KJ851736.1 | - | 3 | 493 | |||
KJ851737.1 | - | 3 | 493 | |||
KJ851738.1 | - | 3 | 493 | |||
KJ851739.1 | - | 3 | 493 | |||
KJ851740.1 | - | 3 | 493 | |||
KJ851741.1 | - | 3 | 493 | |||
KJ851760.1 | - | 4 | 627 | |||
KJ851761.1 | - | 4 | 627 | |||
KJ851762.1 | - | 4 | 627 | |||
KJ851763.1 | - | 4 | 627 | |||
KJ851764.1 | - | 4 | 627 | |||
KJ851765.1 | - | 4 | 627 | |||
KJ851766.1 | - | 4 | 627 | |||
KJ851767.1 | - | 4 | 627 | |||
KJ851768.1 | - | 4 | 627 | |||
KJ851769.1 | - | 4 | 627 | |||
KJ851770.1 | - | 4 | 627 | |||
KJ851771.1 | - | 4 | 627 | |||
KJ851772.1 | - | 4 | 627 | |||
KJ851773.1 | - | 4 | 627 | |||
KJ851774.1 | - | 4 | 627 | |||
KJ851775.1 | - | 4 | 627 | |||
KJ851776.1 | - | 4 | 627 | |||
KJ851777.1 | - | 4 | 627 | |||
KJ851796.1 | - | 3 | 493 | |||
KJ851797.1 | - | 4 | 627 |