Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320395.1 | NP_001307324.1 | 4 | 1012 | ||
NM_001320397.1 | NP_001307326.1 | 5 | 969 | |||
NM_001320398.1 | NP_001307327.1 | 5 | 1065 | |||
NM_001320399.1 | NP_001307328.1 | 6 | 1194 | |||
NM_001320400.1 | NP_001307329.1 | 3 | 937 | |||
NM_001320401.1 | NP_001307330.1 | 4 | 993 | |||
NM_020357.2 | NP_065090.1 | 5 | 1068 | |||
NR_135223.1 | 5 | 1016 | ||||
NR_135224.1 | 4 | 853 | ||||
NR_135225.1 | 5 | 1013 | ||||
NR_135226.1 | 3 | 885 | ||||
NR_135227.1 | 5 | 1269 | ||||
NR_135228.1 | 4 | 960 | ||||
NR_135229.1 | 5 | 1197 | ||||
NR_135230.1 | 4 | 941 | ||||
NR_135231.1 | 3 | 778 | ||||
NR_135232.1 | 6 | 1111 | ||||
XM_017006922.1 | XP_016862411.1 | 4 | 923 | |||
GenBank mRNA | AB037675.1 | - | 5 | 964 | ||
AK025551.1 | - | 5 | 908 | |||
AK098760.1 | - | 5 | 947 | |||
BC013916.1 | - | 4 | 920 | 3929908 | ||
BC022001.2 | - | 5 | 966 | 4823990 | ||
BC107796.1 | - | 5 | 910 | |||
BX647886.1 | - | 1 | 1654 | |||
CR627371.1 | - | 4 | 754 |