Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145056.1 | NP_001138528.1 | 14 | 1255 | ||
NM_020428.3 | NP_065161.3 | 14 | 1314 | |||
XM_005259997.1 | XP_005260054.1 | 14 | 1314 | |||
XM_005259999.1 | XP_005260056.1 | 14 | 1255 | |||
GenBank mRNA | AK027519.1 | - | 14 | 1199 | ||
AK075512.1 | - | 3 | 216 | |||
AK126550.1 | - | 2 | 65 | |||
AK309801.1 | - | 11 | 1255 | |||
AK314582.1 | - | 14 | 1256 | |||
AL832517.1 | - | 4 | 360 | |||
AL832978.1 | - | 11 | 998 | |||
BC010617.2 | - | 11 | 1005 | |||
BC040556.1 | - | 14 | 1256 | 5268313 | ||
BC042692.1 | - | 14 | 1256 | |||
FN668568.1 | - | 14 | 1314 |