Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 6 | 754 | ||
NM_020640.3 | NP_065691.2 | 6 | 736 | |||
XM_011512912.1 | XP_011511214.1 | 6 | 705 | |||
XM_011512914.1 | XP_011511216.1 | 6 | 744 | |||
XM_011512915.2 | XP_011511217.1 | 7 | 1266 | |||
XM_011512916.2 | XP_011511218.1 | 6 | 1194 | |||
XM_017006633.1 | XP_016862122.1 | 5 | 621 | |||
XM_017006634.1 | XP_016862123.1 | 6 | 710 | |||
XM_017006635.1 | XP_016862124.1 | 7 | 1266 | |||
GenBank mRNA | AF292100.2 | - | 6 | 653 | ||
AF456425.1 | - | 6 | 639 | |||
AF456426.1 | - | 6 | 639 | |||
AK025764.1 | - | 4 | 299 | |||
AK056335.1 | - | 6 | 669 | |||
AK297328.1 | - | 7 | 553 | |||
AK314480.1 | - | 6 | 653 | |||
BC009478.2 | - | 6 | 658 | 3939758 | ||
BC013163.1 | - | 5 | 609 | |||
BI828039.1 | - | 6 | 694 |