Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320334.1 | NP_001307263.1 | 3 | 404 | ||
NM_020696.3 | NP_065747.1 | 3 | 349 | |||
GenBank mRNA | AB032969.1 | - | 2 | 156 | ||
AK289549.1 | - | 3 | 321 | |||
BC008468.1 | - | 3 | 324 | 4277104 | ||
BC016790.1 | - | 3 | 283 | 4102473 | ||
BC070360.1 | - | 3 | 279 | |||
CD049061.1 | - | 3 | 346 | |||
DA183520.1 | - | 3 | 342 | |||
DA192476.1 | - | 3 | 324 | |||
DB238094.1 | - | 3 | 360 |