Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020817.1 | NP_065868.1 | 1 | 219 | ||
XM_006713715.1 | XP_006713778.1 | 1 | 200 | |||
XM_011513052.2 | XP_011511354.1 | 1 | 161 | |||
XM_011513053.1 | XP_011511355.1 | 1 | 155 | |||
XM_011513054.2 | XP_011511356.1 | 1 | 41 | |||
XM_017006956.1 | XP_016862445.1 | 1 | 262 | |||
GenBank mRNA | AF509494.1 | - | 1 | 219 | ||
AK125304.1 | - | 1 | 123 | |||
AK295855.1 | - | 1 | 145 | |||
AK302488.1 | - | 1 | 155 | |||
BC150592.1 | - | 1 | 200 |