Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020817.1 | NP_065868.1 | 17 | 2967 | ||
XM_005247681.3 | XP_005247738.1 | 11 | 1952 | |||
XM_006713715.1 | XP_006713778.1 | 16 | 2909 | |||
XM_011513051.1 | XP_011511353.1 | 16 | 3416 | |||
XM_011513052.2 | XP_011511354.1 | 15 | 2723 | |||
XM_011513053.1 | XP_011511355.1 | 16 | 2859 | |||
XM_011513054.2 | XP_011511356.1 | 16 | 2695 | |||
XM_017006956.1 | XP_016862445.1 | 15 | 2829 | |||
XM_017006957.1 | XP_016862446.1 | 16 | 3514 | |||
XM_017006958.1 | XP_016862447.1 | 16 | 3361 | |||
XM_017006959.1 | XP_016862448.1 | 14 | 2455 | |||
GenBank mRNA | AB037828.1 | - | 13 | 2245 | ||
AF509494.1 | - | 17 | 2967 | |||
BC150592.1 | - | 15 | 2967 |