Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 1 | 1105 | ||
XM_011532158.2 | XP_011530460.1 | 2 | 903 | |||
XM_011532159.1 | XP_011530461.1 | 1 | 1092 | |||
GenBank mRNA | AB055660.1 | - | 1 | 1106 | ||
AK055294.1 | - | 1 | 339 |