Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 6 | 4756 | ||
XM_005263163.4 | XP_005263220.1 | 5 | 9210 | |||
XM_005263164.4 | XP_005263221.1 | 5 | 3768 | |||
XM_005263165.4 | XP_005263222.1 | 5 | 5194 | |||
XM_006714282.3 | XP_006714345.1 | 6 | 3911 | |||
XM_011532157.2 | XP_011530459.2 | 5 | 4598 | |||
XM_011532158.2 | XP_011530460.1 | 7 | 4557 | |||
XM_011532159.1 | XP_011530461.1 | 6 | 4746 | |||
XM_011532160.1 | XP_011530462.1 | 4 | 3758 | |||
XM_011532161.2 | XP_011530463.1 | 5 | 3936 | |||
XM_017008488.1 | XP_016863977.1 | 6 | 3914 | |||
XM_017008489.1 | XP_016863978.1 | 6 | 3815 | |||
GenBank mRNA | AB040914.1 | - | 2 | 1955 | ||
AB055660.1 | - | 6 | 4757 | |||
AK128677.1 | - | 2 | 3258 | |||
BC007291.1 | - | 2 | 117 |