Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292017.1 | NP_001278946.1 | 6 | 1370 | ||
NM_021204.4 | NP_067027.1 | 6 | 1399 | |||
NR_120457.1 | 5 | 1167 | ||||
GenBank mRNA | AF113125.1 | - | 6 | 1268 | ||
AF177286.1 | - | 4 | 835 | |||
AK022656.1 | - | 6 | 1265 | |||
AL110167.1 | - | 2 | 500 | |||
BC001317.1 | - | 5 | 985 | |||
BC065815.1 | - | 6 | 1237 | 6526329 |