Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282719.1 | NP_001269648.1 | 4 | 483 | ||
NM_001282720.1 | NP_001269649.1 | 4 | 465 | |||
NM_001282721.1 | NP_001269650.1 | 4 | 453 | |||
NM_001282722.1 | NP_001269651.1 | 3 | 309 | |||
NM_021925.3 | NP_068744.1 | 5 | 609 | |||
XM_011533014.2 | XP_011531316.1 | 5 | 636 | |||
GenBank mRNA | AK025473.1 | - | 5 | 547 | ||
AK299555.1 | - | 4 | 454 | |||
AK316162.1 | - | 4 | 412 | |||
AK316204.1 | - | 3 | 280 | |||
AK316310.1 | - | 4 | 436 | |||
BC017473.1 | - | 5 | 566 | 3611020 |