Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEFSEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021937.4 | NP_068756.2 | 5 | 1142 | ||
XM_005247696.2 | XP_005247753.1 | 4 | 978 | |||
XM_011513066.2 | XP_011511368.1 | 3 | 590 | |||
XM_017006975.1 | XP_016862464.1 | 5 | 1099 | |||
XM_017006976.1 | XP_016862465.1 | 5 | 1099 | |||
XM_017006977.1 | XP_016862466.1 | 5 | 1099 | |||
XM_017006978.1 | XP_016862467.1 | 6 | 922 | |||
XM_017006979.1 | XP_016862468.1 | 5 | 1411 | |||
XM_017006980.1 | XP_016862469.1 | 5 | 1143 | |||
XM_017006981.1 | XP_016862470.1 | 2 | 937 | |||
GenBank mRNA | BC007933.2 | - | 5 | 1020 | 4120858 |