Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003007.4 | NP_002998.1 | 2 | 255 | ||
GenBank mRNA | BC005229.1 | - | 2 | 186 | ||
BC007096.1 | - | 2 | 186 | 4251523 | ||
BC055416.1 | - | 2 | 186 | 6668871 | ||
BP325874.1 | - | 2 | 255 | |||
BT007177.1 | - | 2 | 165 | |||
DA874748.1 | - | 2 | 222 | |||
J04440.1 | - | 2 | 171 |