Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022168.3 | NP_071451.2 | 5 | 1474 | ||
GenBank mRNA | AF095844.1 | - | 5 | 1241 | ||
AK292941.1 | - | 6 | 1388 | |||
AK314611.1 | - | 5 | 1469 | |||
AY017378.1 | - | 5 | 1227 | |||
BC078180.1 | - | 5 | 1296 | |||
BC111750.1 | - | 5 | 1141 | 40008600 |