Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022346.4 | NP_071741.2 | 3 | 709 | ||
NR_073124.1 | 3 | 709 | ||||
XM_011513876.2 | XP_011512178.1 | 3 | 593 | |||
XM_017008543.1 | XP_016864032.1 | 3 | 593 | |||
XM_017008544.1 | XP_016864033.1 | 3 | 543 | |||
GenBank mRNA | AB013299.1 | - | 3 | 583 | ||
AF235023.1 | - | 3 | 512 | |||
AF331796.1 | - | 3 | 620 | |||
AK022512.1 | - | 3 | 540 | |||
AK023147.1 | - | 2 | 290 | |||
BC068467.1 | - | 3 | 526 | 4821684 | ||
BC101476.1 | - | 3 | 497 | 8068982 | ||
BX474610.1 | - | 3 | 536 |