Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289407.1 | NP_001276336.1 | 3 | 516 | ||
NM_022736.2 | NP_073573.2 | 3 | 547 | |||
GenBank mRNA | AB014732.1 | - | 3 | 379 | ||
AK024215.1 | - | 3 | 350 | |||
AK297593.1 | - | 3 | 547 | |||
AK301680.1 | - | 3 | 286 | |||
BC042197.1 | - | 3 | 359 | 5737259 |