Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135095.1 | NP_001128567.1 | 6 | 674 | ||
NM_022763.3 | NP_073600.3 | 6 | 747 | |||
XM_017007064.1 | XP_016862553.1 | 6 | 727 | |||
GenBank mRNA | AB098597.1 | - | 5 | 632 | ||
AK127826.1 | - | 6 | 727 | |||
AK223599.1 | - | 6 | 778 | |||
AK314478.1 | - | 6 | 674 | |||
AY358367.1 | - | 6 | 716 | |||
BC039297.1 | - | 6 | 638 | 5297602 | ||
BX648340.1 | - | 6 | 669 |