Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135095.1 | NP_001128567.1 | 9 | 1149 | ||
NM_022763.3 | NP_073600.3 | 9 | 1222 | |||
XM_017007062.1 | XP_016862551.1 | 8 | 920 | |||
XM_017007063.1 | XP_016862552.1 | 4 | 406 | |||
XM_017007064.1 | XP_016862553.1 | 9 | 1202 | |||
GenBank mRNA | AB098597.1 | - | 8 | 1107 | ||
AK127826.1 | - | 9 | 1202 | |||
AK223599.1 | - | 9 | 1253 | |||
AK314478.1 | - | 9 | 1149 | |||
AY358146.1 | - | 2 | 95 | |||
AY358367.1 | - | 9 | 1191 | |||
BC039297.1 | - | 9 | 1113 | 5297602 | ||
BX648340.1 | - | 9 | 1144 |