Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022780.3 | NP_073617.1 | 9 | 3404 | ||
GenBank mRNA | AK025448.1 | - | 5 | 2485 | ||
AL050139.1 | - | 1 | 752 | |||
AL832022.1 | - | 4 | 2320 | |||
BC012165.1 | - | 9 | 3125 | 4636136 | ||
BC047668.1 | - | 10 | 2239 | 6066798 |