Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK058094.1 | - | 8 | 1391 | ||
AK298119.1 | - | 6 | 524 | |||
BC005913.1 | - | 6 | 741 | 4043169 | ||
J03517.1 | - | 6 | 745 | |||
J03553.1 | - | 6 | 892 |