Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026052.1 | 2 | 747 | |||
GenBank mRNA | AK000827.1 | - | 2 | 684 | ||
AK311347.1 | - | 2 | 684 | |||
BC000766.2 | - | 2 | 656 | |||
BC001952.1 | - | 2 | 656 | |||
BC043401.1 | - | 2 | 686 | |||
BC067232.1 | - | 2 | 691 | |||
BC098570.1 | - | 1 | 244 | |||
BC119702.1 | - | 1 | 109 | |||
BC119703.1 | - | 1 | 108 |