Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024038.2 | NP_076943.1 | 2, 3 | 479 | ||
GenBank mRNA | AF061732.1 | - | 1, 2 | 55 | ||
AK027588.1 | - | 2, 3 | 374 | |||
AK315754.1 | - | 2, 3 | 445 | |||
BC000216.2 | - | 2, 3 | 479 | 2961319 | ||
BC004533.2 | - | 2, 3 | 424 | 3139656 | ||
BC015347.1 | - | 2, 3 | 438 | 4156774 |