Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320147.1 | NP_001307076.1 | 5 | 855 | ||
NM_024046.4 | NP_076951.2 | 5 | 855 | |||
GenBank mRNA | AK074856.1 | - | 5 | 569 | ||
AK074899.1 | - | 5 | 539 | |||
AK075294.1 | - | 5 | 550 | |||
AK297258.1 | - | 5 | 501 | |||
AK297437.1 | - | 5 | 495 | |||
AK299953.1 | - | 5 | 498 | |||
AK315525.1 | - | 5 | 588 | |||
AL136697.1 | - | 5 | 538 | |||
AL833943.1 | - | 4 | 752 | |||
BC000497.2 | - | 5 | 524 | 2820734 | ||
BC001921.1 | - | 5 | 524 | 2820734 | ||
BC005828.2 | - | 5 | 524 | 2820734 | ||
BC017363.2 | - | 5 | 526 | 3940018 | ||
BC019256.1 | - | 5 | 519 | 3502894 | ||
BX647074.1 | - | 5 | 536 | |||
CR533508.1 | - | 4 | 376 |