Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 13 | 2363 | ||
NM_024085.4 | NP_076990.4 | 12 | 2311 | |||
NR_104255.1 | 13 | 2304 | ||||
GenBank mRNA | AK021732.1 | - | 3 | 449 | ||
AK025822.1 | - | 7 | 1665 | |||
AK027448.1 | - | 2 | 288 | |||
AK056595.1 | - | 13 | 2304 | |||
AK289755.1 | - | 13 | 2258 | |||
AK302521.1 | - | 12 | 2202 | |||
AL833865.1 | - | 6 | 1560 | |||
BC001098.2 | - | 5 | 838 | |||
BC021202.2 | - | 4 | 669 | |||
BC065534.1 | - | 13 | 2242 | 5787445 | ||
BX537984.1 | - | 12 | 2232 | |||
BX538192.1 | - | 12 | 2206 | |||
BX538198.1 | - | 13 | 2140 |