Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003021.3 | NP_003012.1 | 12 | 1145 | ||
XM_011528178.2 | XP_011526480.1 | 12 | 1199 | |||
GenBank mRNA | AF368279.1 | - | 11 | 1019 | ||
AJ133129.1 | - | 12 | 1040 | |||
AJ223828.1 | - | 11 | 1039 | |||
AL050156.1 | - | 12 | 1107 | |||
BC000390.2 | - | 12 | 1048 | 2821268 | ||
BC002989.1 | - | 12 | 1048 | 2821268 | ||
BC005165.2 | - | 12 | 1042 | 3532317 | ||
BC008885.2 | - | 12 | 1042 | 4302652 |