Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282495.1 | NP_001269424.1 | 4, 5 | 534 | ||
NM_001282496.1 | NP_001269425.1 | 3, 4 | 566 | |||
NM_001282497.1 | NP_001269426.1 | 3, 4 | 608 | |||
NM_001282499.1 | NP_001269428.1 | 3, 4 | 608 | |||
NM_001321800.1 | NP_001308729.1 | 3, 4 | 472 | |||
NM_001321801.1 | NP_001308730.1 | 3, 4 | 472 | |||
NM_001321802.1 | NP_001308731.1 | 3, 4 | 608 | |||
NM_001321803.1 | NP_001308732.1 | 3, 4 | 472 | |||
NM_001321805.1 | NP_001308734.1 | 3, 4 | 608 | |||
NM_024325.5 | NP_077301.4 | 3, 4 | 608 | |||
XM_017028062.1 | XP_016883551.1 | 3, 4 | 449 | |||
GenBank mRNA | AK096911.1 | - | 3, 4 | 609 | ||
BC002897.2 | - | 3, 4 | 398 | |||
BC065009.1 | - | 4, 5 | 531 | 6166526 | ||
CD702168.1 | - | 3, 4 | 360 | |||
DB217060.1 | - | 3, 4 | 472 | |||
DR002765.1 | - | 3, 4 | 586 |