Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024656.3 | NP_078932.2 | 5, 6 | 959 | ||
XM_005260080.4 | XP_005260137.1 | 5, 6 | 605 | |||
XM_011528297.2 | XP_011526599.1 | 5, 6 | 597 | |||
XM_011528298.2 | XP_011526600.1 | 5, 6 | 698 | |||
XM_011528299.2 | XP_011526601.1 | 3, 4 | 472 | |||
GenBank mRNA | AK074941.1 | - | 5, 6 | 963 | ||
AK075541.1 | - | 5, 6 | 851 | |||
BC108308.1 | - | 5, 6 | 914 | 5138787 |