Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125487
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
7
1036
NM_001164629.3
NP_001158101.1
7
1021
NM_001284233.1
NP_001271162.1
5
845
NM_001284234.1
NP_001271163.1
5
736
NM_001284235.1
NP_001271164.1
7
1003
NM_001284238.1
NP_001271167.1
9
1235
NM_024659.4
NP_078935.2
7
1003
XM_005263774.2
XP_005263831.1
8
1120
XM_005263782.2
XP_005263839.1
8
1136
XM_005263784.2
XP_005263841.1
6
995
XM_011511843.2
XP_011510145.1
8
1169
XM_011511855.2
XP_011510157.1
6
1113
XM_011511856.2
XP_011510158.1
6
895
XM_017004917.1
XP_016860406.1
12
1791
XM_017004918.1
XP_016860407.1
10
1383
XM_017004919.1
XP_016860408.1
8
1269
XM_017004920.1
XP_016860409.1
10
4425
XM_017004921.1
XP_016860410.1
9
1218
XM_017004922.1
XP_016860411.1
9
1395
XM_017004923.1
XP_016860412.1
8
1183
XM_017004924.1
XP_016860413.1
10
1379
XM_017004925.1
XP_016860414.1
7
1041
XM_017004926.1
XP_016860415.1
5
972
XM_017004927.1
XP_016860416.1
7
1003
XM_017004928.1
XP_016860417.1
9
1236
XM_017004929.1
XP_016860418.1
7
973
XM_017004930.1
XP_016860419.1
7
1624
XM_017004931.1
XP_016860420.1
7
1037
XM_017004932.1
XP_016860421.1
7
1007
XM_017004933.1
XP_016860422.1
7
989
XM_017004934.1
XP_016860423.1
8
4196
XM_017004935.1
XP_016860424.1
9
2066
XM_017004936.1
XP_016860425.1
8
1243
XM_017004937.1
XP_016860426.1
6
1124
XM_017004938.1
XP_016860427.1
6
1122
XM_017004939.1
XP_016860428.1
8
1172
XM_017004940.1
XP_016860429.1
5
973
XM_017004941.1
XP_016860430.1
5
972
XM_017004942.1
XP_016860431.1
7
1019
XM_017004943.1
XP_016860432.1
7
1685
XM_017004944.1
XP_016860433.1
7
1018
XM_017004945.1
XP_016860434.1
9
1236
XM_017004946.1
XP_016860435.1
7
973
XM_017004947.1
XP_016860436.1
7
1656
XM_017004948.1
XP_016860437.1
8
4196
GenBank mRNA
AB201269.1
-
4
757
AB597184.1
-
4
757
AK021815.1
-
1
212
AK091684.1
-
5
849
AK291357.1
-
7
951
AY281366.1
-
7
1036
BC017741.2
-
8
1043
4390876
BC061699.1
-
7
943
4608518
BX431825.2
-
1
159
BX647436.1
-
5
718
BX647773.1
-
9
1174
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline