Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256964.1 | NP_001243893.1 | 4 | 1066 | ||
NM_001256965.2 | NP_001243894.1 | 4 | 1050 | |||
NM_001256966.2 | NP_001243895.1 | 4 | 945 | |||
NM_001256967.2 | NP_001243896.1 | 4 | 940 | |||
NM_001256968.2 | NP_001243897.1 | 4 | 837 | |||
NM_001256969.2 | NP_001243898.1 | 4 | 832 | |||
NM_024661.4 | NP_078937.3 | 4 | 958 | |||
XM_011534113.2 | XP_011532415.1 | 5 | 1393 | |||
GenBank mRNA | AK022498.1 | - | 4 | 837 | ||
BC004341.1 | - | 2 | 697 | |||
BC011993.1 | - | 3 | 948 | 3857480 | ||
BQ427785.1 | - | 3 | 842 |