Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1XR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321193.1 | NP_001308122.1 | 4 | 347 | ||
NM_001321194.1 | NP_001308123.1 | 5 | 527 | |||
NM_001321195.1 | NP_001308124.1 | 3 | 244 | |||
NM_024665.5 | NP_078941.2 | 4 | 408 | |||
XM_005247775.2 | XP_005247832.1 | 3 | 363 | |||
XM_006713746.1 | XP_006713809.1 | 5 | 417 | |||
XM_011513141.1 | XP_011511443.1 | 5 | 351 | |||
XM_011513142.2 | XP_011511444.1 | 4 | 332 | |||
XM_011513143.2 | XP_011511445.1 | 3 | 258 | |||
XM_017007185.1 | XP_016862674.1 | 4 | 345 | |||
GenBank mRNA | AF314544.1 | - | 3 | 207 | ||
AK022956.1 | - | 4 | 307 | |||
AK296946.1 | - | 3 | 142 | |||
BC060320.1 | - | 3 | 261 | |||
BC113421.1 | - | 3 | 150 | 8322473 | ||
BX648935.1 | - | 4 | 342 | |||
DA159105.1 | - | 3 | 193 | |||
DA460721.1 | - | 2 | 169 | |||
DB150466.1 | - | 5 | 426 | |||
DB459301.1 | - | 4 | 409 |