Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1XR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321193.1 | NP_001308122.1 | 16 | 2973 | ||
NM_001321194.1 | NP_001308123.1 | 17 | 3153 | |||
NM_001321195.1 | NP_001308124.1 | 15 | 2870 | |||
NM_024665.5 | NP_078941.2 | 16 | 3034 | |||
XM_005247775.2 | XP_005247832.1 | 15 | 2989 | |||
XM_006713746.1 | XP_006713809.1 | 17 | 3043 | |||
XM_011513141.1 | XP_011511443.1 | 17 | 2977 | |||
XM_011513142.2 | XP_011511444.1 | 16 | 2958 | |||
XM_011513143.2 | XP_011511445.1 | 15 | 2884 | |||
XM_017007185.1 | XP_016862674.1 | 16 | 2971 | |||
GenBank mRNA | AF268193.1 | - | 16 | 2865 | ||
BX648080.1 | - | 2 | 2646 | |||
BX648935.1 | - | 16 | 2970 |