Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBBX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199201.1 | NP_001186130.1 | 19 | 2247 | ||
XM_011513144.1 | XP_011511446.1 | 19 | 2247 | |||
XM_011513145.2 | XP_011511447.2 | 18 | 2273 | |||
XM_011513148.2 | XP_011511450.2 | 19 | 2244 | |||
XM_011513149.2 | XP_011511451.2 | 20 | 2364 | |||
XM_011513151.2 | XP_011511453.2 | 21 | 2553 | |||
XM_011513152.2 | XP_011511454.2 | 13 | 1674 | |||
XM_011513153.2 | XP_011511455.2 | 10 | 1346 | |||
XM_017007186.1 | XP_016862675.1 | 19 | 2247 | |||
XM_017007187.1 | XP_016862676.1 | 19 | 2247 | |||
XM_017007190.1 | XP_016862679.1 | 19 | 2247 | |||
GenBank mRNA | AK026702.1 | - | 10 | 1096 | ||
BC144444.1 | - | 18 | 2194 | 9052969 |