Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 2090 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 2051 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 2048 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 1991 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 1952 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 1949 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 1988 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 2007 | |||
NM_001291745.1 | NP_001278674.1 | 4 | 539 | |||
NM_024691.3 | NP_078967.3 | 5 | 2087 | |||
GenBank mRNA | AB209371.1 | - | 3 | 5336 | ||
AK026886.1 | - | 4 | 1960 | |||
AK302135.1 | - | 3 | 1910 | |||
AK304569.1 | - | 4 | 1996 | |||
AK316156.1 | - | 4 | 2009 | |||
BC008297.1 | - | 5 | 2033 | 3544044 | ||
BC024259.1 | - | 5 | 2033 | 3604215 | ||
BF110540.1 | - | 4 | 294 |
Gene Symbol: ZNF773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304334.1 | NP_001291263.1 | 4 | 2338 | ||
NM_001304336.1 | NP_001291265.1 | 4 | 2334 | |||
NM_198542.2 | NP_940944.1 | 4 | 2341 |