Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 2361 | |||
NM_001098492.1 | NP_001091962.1 | 2322 | ||||
NM_001098493.1 | NP_001091963.1 | 2319 | ||||
NM_001098494.1 | NP_001091964.1 | 2262 | ||||
NM_001098495.1 | NP_001091965.1 | 2223 | ||||
NM_001098496.1 | NP_001091966.1 | 2220 | ||||
NM_001291743.1 | NP_001278672.1 | 2259 | ||||
NM_001291744.1 | NP_001278673.1 | 2278 | ||||
NM_001291745.1 | NP_001278674.1 | 810 | ||||
NM_024691.3 | NP_078967.3 | 2358 | ||||
GenBank mRNA | BC008297.1 | - | 2304 | 3544044 | ||
BC024259.1 | - | 2304 | 3604215 |