Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024753.4 | NP_079029.3 | 4 | 520 | ||
XM_006712761.1 | XP_006712824.1 | 4 | 463 | |||
XM_011511872.2 | XP_011510174.1 | 4 | 463 | |||
XM_017004967.1 | XP_016860456.1 | 4 | 463 | |||
XM_017004969.1 | XP_016860458.1 | 2 | 163 | |||
GenBank mRNA | AB073395.1 | - | 4 | 427 | ||
AK057268.1 | - | 4 | 458 | |||
AK292942.1 | - | 4 | 463 | |||
BC055424.1 | - | 4 | 520 | 4837489 | ||
DA798655.1 | - | 2 | 247 |