Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024753.4 | NP_079029.3 | 26 | 3635 | ||
XM_011511871.2 | XP_011510173.1 | 21 | 6880 | |||
XM_011511872.2 | XP_011510174.1 | 26 | 3609 | |||
XM_017004967.1 | XP_016860456.1 | 26 | 3578 | |||
XM_017004968.1 | XP_016860457.1 | 21 | 5126 | |||
XM_017004969.1 | XP_016860458.1 | 24 | 3295 | |||
GenBank mRNA | AK096451.1 | - | 10 | 1262 | ||
AK292942.1 | - | 26 | 3578 | |||
BC035767.2 | - | 3 | 401 | |||
BC055424.1 | - | 26 | 3635 | 4837489 | ||
BC063579.1 | - | 8 | 979 | |||
BX647903.1 | - | 12 | 1386 |