Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024768.2 | NP_079044.2 | 8 | 2382 | ||
XM_005247781.3 | XP_005247838.1 | 8 | 2410 | |||
XM_011513161.2 | XP_011511463.1 | 8 | 1842 | |||
XM_011513164.2 | XP_011511466.1 | 7 | 2455 | |||
GenBank mRNA | AK022119.1 | - | 6 | 1656 |