Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048166.1 | NP_001041631.1 | 5, 6 | 616 | ||
NM_001282936.1 | NP_001269865.1 | 6, 7 | 825 | |||
NM_001282937.1 | NP_001269866.1 | 6, 7 | 825 | |||
NM_001282938.1 | NP_001269867.1 | 6, 7 | 825 | |||
NM_001282939.1 | NP_001269868.1 | 6, 7 | 690 | |||
NM_003035.2 | NP_003026.2 | 5, 6 | 616 | |||
XM_006710834.3 | XP_006710897.1 | 6, 7 | 690 | |||
XM_011541991.2 | XP_011540293.1 | 6, 7 | 1356 | |||
XM_011541992.2 | XP_011540294.1 | 6, 7 | 1339 | |||
XM_011541994.2 | XP_011540296.1 | 5, 6 | 616 | |||
XM_011541996.2 | XP_011540298.1 | 5, 6 | 616 | |||
XM_011541998.2 | XP_011540300.1 | 5, 6 | 616 | |||
XM_011542001.2 | XP_011540303.1 | 5, 6 | 616 | |||
XM_017002123.1 | XP_016857612.1 | 5, 6 | 616 | |||
XM_017002125.1 | XP_016857614.1 | 5, 6 | 616 | |||
XM_017002126.1 | XP_016857615.1 | 5, 6 | 616 | |||
XM_017002127.1 | XP_016857616.1 | 5, 6 | 616 | |||
GenBank mRNA | AK307519.1 | - | 5, 6 | 578 | ||
BC053615.1 | - | 6, 7 | 658 | |||
BC126223.1 | - | 5, 6 | 692 | 8991939 | ||
BC144089.1 | - | 5, 6 | 692 | 9052609 | ||
CR749851.1 | - | 5, 6 | 592 | |||
DA754072.1 | - | 5, 6 | 543 | |||
M74558.1 | - | 6, 7 | 841 |