Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261392.1 | NP_001248321.1 | 6 | 552 | ||
NM_001261394.1 | NP_001248323.1 | 7 | 630 | |||
NM_024894.3 | NP_079170.2 | 7 | 630 | |||
NR_048552.1 | 6 | 584 | ||||
XM_011510398.2 | XP_011508700.1 | 6 | 497 | |||
XM_011510399.2 | XP_011508701.1 | 5 | 435 | |||
XM_011510400.2 | XP_011508702.1 | 7 | 618 | |||
XM_011510402.2 | XP_011508704.1 | 7 | 618 | |||
XM_017004990.1 | XP_016860479.1 | 5 | 372 | |||
GenBank mRNA | AK024000.1 | - | 6 | 471 | ||
AK024137.1 | - | 7 | 615 | |||
AK290680.1 | - | 7 | 594 | |||
AK297169.1 | - | 6 | 540 | |||
AK299617.1 | - | 6 | 572 | |||
BC005125.2 | - | 7 | 610 | 3841643 | ||
BG390065.1 | - | 2 | 84 |