Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115116.1 | NP_001108588.1 | 6 | 1712 | ||
NM_024933.3 | NP_079209.3 | 7 | 1839 | |||
XM_005264569.4 | XP_005264626.1 | 6 | 2872 | |||
XM_011533114.2 | XP_011531416.1 | 5 | 1383 | |||
GenBank mRNA | AK093479.1 | - | 6 | 1548 | ||
BC035234.1 | - | 7 | 1839 | 5166484 |