Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308164.1 | NP_001295093.1 | 2 | 308 | ||
NM_001308166.1 | NP_001295095.1 | 2 | 308 | |||
NM_024996.5 | NP_079272.4 | 2 | 308 | |||
XM_006713795.1 | XP_006713858.1 | 2 | 339 | |||
GenBank mRNA | AF309777.1 | - | 2 | 308 | ||
AF367998.1 | - | 2 | 308 | |||
AK000780.1 | - | 2 | 308 | |||
AK315031.1 | - | 2 | 247 | |||
BC049210.1 | - | 2 | 272 | 5574223 | ||
KU178816.1 | - | 2 | 201 | |||
KU178817.1 | - | 2 | 201 |