Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWH43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286791.1 | NP_001273720.1 | 7 | 1112 | ||
NM_025087.2 | NP_079363.2 | 7 | 1193 | |||
XM_011513755.1 | XP_011512057.1 | 7 | 1194 | |||
XM_011513756.2 | XP_011512058.1 | 7 | 1225 | |||
XM_011513757.2 | XP_011512059.1 | 7 | 1251 | |||
XM_011513759.1 | XP_011512061.1 | 4 | 590 | |||
GenBank mRNA | AK025164.1 | - | 7 | 1193 | ||
AK300495.1 | - | 7 | 1115 | |||
AK316307.1 | - | 7 | 1085 | |||
BC137388.1 | - | 7 | 1176 | 9021386 |