Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320818.1 | NP_001307747.1 | 2 | 387 | ||
NM_001320819.1 | NP_001307748.1 | 3 | 581 | |||
NM_001320820.1 | NP_001307749.1 | 3 | 778 | |||
NM_025147.4 | NP_079423.1 | 3 | 537 | |||
GenBank mRNA | AK023510.1 | - | 3 | 406 | ||
AK294093.1 | - | 3 | 778 | |||
AK308977.1 | - | 3 | 375 | |||
BC022453.1 | - | 3 | 454 | 4796819 | ||
BU195968.1 | - | 2 | 321 | |||
DC328985.1 | - | 3 | 375 | |||
DC349756.1 | - | 3 | 536 |