Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 2, 3 | 261 | ||
NM_025176.5 | NP_079452.3 | 2, 3 | 261 | |||
XM_011529186.2 | XP_011527488.1 | 2, 3 | 884 | |||
XM_011529187.2 | XP_011527489.1 | 2, 3 | 884 | |||
XM_011529188.1 | XP_011527490.1 | 2, 3 | 261 | |||
XM_011529189.1 | XP_011527491.1 | 2, 3 | 269 | |||
XM_011529190.1 | XP_011527492.1 | 1, 2 | 184 | |||
XM_011529193.1 | XP_011527495.1 | 1, 2 | 184 | |||
XM_017027732.1 | XP_016883221.1 | 3, 4 | 2203 | |||
XM_017027733.1 | XP_016883222.1 | 2, 3 | 210 | |||
GenBank mRNA | AB023197.1 | - | 2, 3 | 247 | ||
AK293991.1 | - | 2, 3 | 547 | |||
AK309298.1 | - | 2, 3 | 261 | |||
BC036380.2 | - | 1, 2 | 199 | 5268454 | ||
DA448124.1 | - | 2, 3 | 261 | |||
DA871028.1 | - | 2, 3 | 260 | |||
EU718622.1 | - | 2, 3 | 247 |