Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134367.3 | NP_001127839.2 | 3, 4 | 558 | ||
NM_001134368.3 | NP_001127840.1 | 3, 4 | 562 | |||
NM_003043.5 | NP_003034.2 | 3, 4 | 562 | |||
NR_103507.2 | 3, 4 | 562 | ||||
XM_006713307.2 | XP_006713370.1 | 2, 3 | 366 | |||
XM_011534030.2 | XP_011532332.1 | 2, 3 | 319 | |||
GenBank mRNA | AB209172.1 | - | 3, 4 | 488 | ||
AK303649.1 | - | 3, 4 | 562 | |||
AK308417.1 | - | 2, 3 | 164 | |||
BC006252.2 | - | 3, 4 | 492 | |||
BC111489.1 | - | 3, 4 | 518 | |||
BC137128.1 | - | 3, 4 | 509 | 9021125 | ||
BC137129.1 | - | 3, 4 | 505 | 9021126 | ||
U09220.1 | - | 1, 2 | 237 | |||
U16120.1 | - | 4, 5 | 613 | |||
Z18956.1 | - | 1, 2 | 256 |