Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025205.4 | NP_079481.2 | 1 | 157 | ||
GenBank mRNA | AF317678.1 | - | 1 | 129 | ||
AF317679.1 | - | 1 | 134 | |||
AF317680.1 | - | 1 | 141 | |||
AF318059.1 | - | 1 | 154 | |||
AF358829.1 | - | 1 | 142 | |||
AK313928.1 | - | 1 | 139 | |||
BC011936.2 | - | 1 | 134 | 3936713 | ||
HY174042.1 | - | 1 | 157 |