Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030923.4 | NP_112185.1 | 2 | 315 | ||
XM_011511950.2 | XP_011510252.1 | 2 | 421 | |||
XM_017005060.1 | XP_016860549.1 | 2 | 402 | |||
XM_017005061.1 | XP_016860550.1 | 2 | 291 | |||
XM_017005062.1 | XP_016860551.1 | 2 | 380 | |||
GenBank mRNA | AL122044.1 | - | 4 | 292 | ||
BC026170.2 | - | 2 | 315 | 4837567 |