Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199140.1 | NP_001186069.1 | 4 | 787 | ||
NM_031445.2 | NP_113633.2 | 4 | 759 | |||
XM_005263804.4 | XP_005263861.1 | 4 | 1255 | |||
XM_011511952.2 | XP_011510254.1 | 3 | 1145 | |||
XM_011511954.2 | XP_011510256.1 | 5 | 1347 | |||
XM_011511955.2 | XP_011510257.1 | 2 | 174 | |||
XM_011511956.2 | XP_011510258.1 | 4 | 1255 | |||
XM_017005063.1 | XP_016860552.1 | 4 | 1255 | |||
GenBank mRNA | AK304145.1 | - | 4 | 787 | ||
BC016181.1 | - | 4 | 759 | 4644159 | ||
BC078174.1 | - | 4 | 769 | 6012672 |