Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130011.1 | NP_001123483.1 | 3 | 278 | ||
NM_031451.4 | NP_113639.4 | 6 | 698 | |||
XM_005259303.3 | XP_005259360.2 | 3 | 268 | |||
GenBank mRNA | AF241268.1 | - | 6 | 697 | ||
AF353396.1 | - | 3 | 368 | |||
AY014285.1 | - | 3 | 250 | |||
AY359077.1 | - | 3 | 246 | |||
BC001861.2 | - | 4 | 445 | 3538494 |