Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146102.1 | NP_001139574.1 | 4 | 531 | ||
NM_001146103.1 | NP_001139575.1 | 4 | 608 | |||
NM_001146104.1 | NP_001139576.1 | 4 | 607 | |||
NM_001146105.1 | NP_001139577.1 | 4 | 426 | |||
NM_001146106.1 | NP_001139578.1 | 4 | 607 | |||
NM_031458.2 | NP_113646.2 | 4 | 713 | |||
XM_005247820.2 | XP_005247877.1 | 4 | 637 | |||
XM_011513218.1 | XP_011511520.1 | 4 | 503 | |||
XM_017007304.1 | XP_016862793.1 | 4 | 521 | |||
XM_017007305.1 | XP_016862794.1 | 4 | 416 | |||
XM_017007306.1 | XP_016862795.1 | 4 | 497 | |||
GenBank mRNA | AF307338.1 | - | 4 | 614 | ||
AF307339.1 | - | 4 | 509 | |||
AK292959.1 | - | 4 | 386 | |||
AK299107.1 | - | 4 | 500 | |||
AK313494.1 | - | 4 | 500 | |||
BC017463.1 | - | 4 | 575 | |||
BC039580.1 | - | 3 | 384 | 5416494 | ||
DC368112.1 | - | 4 | 426 |